Stampa…

2,2'-Dithiodipropionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D0946-25G D0946-5G
TCIAD0946-25GEA 408 EUR
TCIAD0946-25G TCIAD0946-5G
2,2'-Dithiodipropionic acid ≥98.0% (by GC, titration analysis)
2,2'-Dithiodipropionic acid
Formula: C₆H₁₀O₄S₂
Peso molecolare: 210,26 g/mol
MDL Number: MFCD00059649
CAS Number: 4775-93-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR