Stampa…

Di(ɑ-phenylethyl) sulphide (DL and meso mixture)

Fornitore: TCI EUROPE

Synonyms: 1-Phenylethyl Sulphide

D0889-25G D0889-5G
TCIAD0889-25GEA 367 EUR
TCIAD0889-25G TCIAD0889-5G
Di(ɑ-phenylethyl) sulphide (DL and meso mixture)
Di(alpha-phenylethyl) sulphide (DL and meso mixture)
Formula: C₁₆H₁₈S
Peso molecolare: 242,38 g/mol
MDL Number: MFCD00026342
CAS Number: 838-59-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white clear liquid
Purity (GC) min. 55.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR