Stampa…

2,2-Dimethyl-1,3-dichloropropane ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3-Dichloro-2,2-dimethylpropane, 1,3-Dichloroneopentane

D0870-25G D0870-5G
TCIAD0870-25GEA 217 EUR
TCIAD0870-25G TCIAD0870-5G
2,2-Dimethyl-1,3-dichloropropane ≥95.0% (by GC)
2,2-Dimethyl-1,3-dichloropropane
Formula: C₅H₁₀Cl₂
Peso molecolare: 141,04 g/mol
MDL Number: MFCD00039363
CAS Number: 29559-55-5
UN: 1993
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR