Stampa…

4,4'-Dinitrodiphenyl ether ≥99.0% (by GC)

Fornitore: TCI
Attenzione

D0832-1G D0832-10G
TCIAD0832-1GEA 16 EUR
TCIAD0832-1G TCIAD0832-10G
4,4'-Dinitrodiphenyl ether ≥99.0% (by GC)
4,4'-Dinitrodiphenyl ether
Formula: C₁₂H₈N₂O₅
MDL Number: MFCD00059182
CAS Number: 101-63-3
EINECS: 202-961-3

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 99.0 %
Melting point 142.0 to 147.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR