Stampa…

Sodium-2,4-dinitrobenzenesulphonate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

D0821-25G D0821-500G
TCIAD0821-25GEA 40 EUR
TCIAD0821-25G TCIAD0821-500G
Sodium-2,4-dinitrobenzenesulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-2,4-dinitrobenzenesulphonate
Formula: C₆H₃N₂NaO₇S
MDL Number: MFCD00007471
CAS Number: 885-62-1
EINECS: 212-943-7
UN: 1325
ADR: 4.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 % (calcd.on dried substance)
Solubility in Water almost transparency
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR