Butopyronoxyl ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylic Acid Butyl Ester, Indalone, Butyl 3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylate
D0550-25G
D0550-500G
TCIAD0550-25GEA
49
EUR
TCIAD0550-25G
TCIAD0550-500G
Butopyronoxyl ≥95.0% (by HPLC)
Butopyronoxyl
Formula:
C₁₂H₁₈O₄ Peso molecolare: 226,27 g/mol Flash Pt: 157°C |
MDL Number:
MFCD00006574 CAS Number: 532-34-3 EINECS: 208-535-3 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (HPLC) | min. 95.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...