Stampa…

Diethylammonium diethyldithiocarbamate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Diethyldithiocarbamic Acid Diethylammonium Salt, Diethylammonium diethylcarbamodithioate

D0488-25G D0488-500G
TCIAD0488-25GEA 68 EUR
TCIAD0488-25G TCIAD0488-500G
Diethylammonium diethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Diethylammonium diethyldithiocarbamate
Formula: C₅H₁₁NS₂
Peso molecolare: 222,41 g/mol
MDL Number: MFCD00013146
CAS Number: 1518-58-7
EINECS: 216-175-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99b1500a922016c30988b919527debeac53f37ae&vid=99b1500a922016c30988b919527debeac53f37ae&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 82.0 to 84.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR