Copper(II) diethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Diethyldithiocarbamic Acid Copper(II) Salt
D0487-25G
D0487-5G
TCIAD0487-25GEA
98
EUR
TCIAD0487-25G
TCIAD0487-5G
Copper(II) diethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Copper(II) diethyldithiocarbamate
Formula:
C₅H₁₁NS₂ Peso molecolare: 360,07 g/mol |
MDL Number:
MFCD00070506 CAS Number: 13681-87-3 |
Risultati del test di specificazione
Appearance | Red to Dark blue to Black powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Melting point | 196.0 to 201.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...