Stampa…

Di-tert-dodecyl disulphide (mixture of isomers)

Fornitore: TCI EUROPE

Synonyms: tert-Dodecyl Disulphide

D0447-25G D0447-250G
TCIAD0447-25GEA 31 EUR
TCIAD0447-25G TCIAD0447-250G
Di-tert-dodecyl disulphide (mixture of isomers)
Di-tert-dodecyl disulphide (mixture of isomers)
Formula: C₂₄H₅₀S₂
Peso molecolare: 402,78 g/mol
Flash Pt: 118°C
MDL Number: MFCD00059944
CAS Number: 27458-90-8
EINECS: 248-468-7

Visualizza i codici

Risultati del test di specificazione

Purity (GC) many component mixture
Appearance Colorless to Light orange to Yellow clear liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR