Fluorescein chloride ≥95.0% (by HPLC) reagent for amines
Fornitore: TCI
Synonyms:
3',6'-Dichloro-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 3',6'-Dichlorofluoran
Formula:
C₂₀H₁₀Cl₂O₃ |
MDL Number:
MFCD00037841 CAS Number: 630-88-6 EINECS: 211-147-7 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Argentmetric Titration) | min. 93.0 % |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...