Stampa…

Dibutyltin maleate (so called) (so called) for PVC stabilizer

Fornitore: TCI
Pericolo

D0304-25G D0304-500G
TCIAD0304-25GEA 16 EUR
TCIAD0304-25G TCIAD0304-500G
Dibutyltin maleate (so called) (so called) for PVC stabilizer
Dibutyltin maleate (so called)
Formula: C₁₂H₂₀O₄Sn
Peso molecolare: 347.00 g/mol
Flash Pt: 132°C
MDL Number: MFCD00014120
CAS Number: 78-04-6
EINECS: 201-077-5
UN: 2788
ADR: 6.1 II

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Ignition residue (as SnO2) 24.0 to 28.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR