2,5-Di-tert-amylhydroquinone ≥93.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,5-Di-tert-pentylhydroquinone
D0299-25G
D0299-500G
TCIAD0299-25GEA
21
EUR
TCIAD0299-25G
TCIAD0299-500G
2,5-Di-tert-amylhydroquinone ≥93.0% (by GC)
2,5-Di-tert-amylhydroquinone
Formula:
C₁₆H₂₆O₂ Peso molecolare: 250,38 g/mol |
MDL Number:
MFCD00027029 CAS Number: 79-74-3 EINECS: 201-222-2 Merck Index: 14,03304 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 93.0 % |
Melting point | 178.0 to 181.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...