Stampa…

4-(Dimethylamino)stilbene ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

D0255-5G D0255-1G
TCIAD0255-5GEA 96 EUR
TCIAD0255-5G TCIAD0255-1G
4-(Dimethylamino)stilbene ≥99.0% (by titrimetric analysis)
4-(Dimethylamino)stilbene
Formula: C₁₆H₁₇N
Peso molecolare: 223,32 g/mol
MDL Number: MFCD00017151
CAS Number: 1145-73-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystaline
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 148.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR