Stampa…

Zinc(II) dibutyldithiocarbamate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Dibutyldithiocarbamic Acid Zinc(II) Salt

D0227-25G D0227-500G
TCIAD0227-25GEA 17 EUR
TCIAD0227-25G TCIAD0227-500G
Zinc(II) dibutyldithiocarbamate ≥98.0% (by titrimetric analysis)
Zinc(II) dibutyldithiocarbamate
Formula: C₉H₁₉NS₂
Peso molecolare: 474.12 g/mol
MDL Number: MFCD00067274
CAS Number: 136-23-2
EINECS: 205-232-8
UN: 3077
ADR: 9 III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C
Solubility in Toluene faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR