Stampa…

Phenoxybenzamine hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dibenzyline Hydrochloride, N-(2-Chloroethyl)-N-(1-methyl-2-phenoxyethyl)benzylamine Hydrochloride, N-Phenoxyisopropyl-N-benzyl-2-chloroethylamine Hydrochloride, N-Benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine hydrochloride

D0158-1G D0158-25G
TCIAD0158-1GEA 37 EUR
TCIAD0158-1G TCIAD0158-25G
Phenoxybenzamine hydrochloride ≥98.0% (by titrimetric analysis)
Phenoxybenzamine hydrochloride
Formula: C₁₈H₂₂ClNO
Peso molecolare: 340,29 g/mol
MDL Number: MFCD00055152
CAS Number: 63-92-3
EINECS: 200-569-7
Merck Index: 14,07256

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR