Stampa…

Argento dietilditiocarbamato ACS

Fornitore: Thermo Fisher Scientific
Attenzione

11898.09 11898.18
11898.09EA 163 EUR
11898.09 11898.18
Argento dietilditiocarbamato ACS
Argento dietilditiocarbammato

CAS No.: 1470-61-7 Molecular Formula: C5H10AgNS2 Formula Weight: 256.14 Physical Form: Powder Sensitivity: Light Sensitive Appearance: Yellow to pale brown Melting Point: 172-175° MDL No.: MFCD00004929

Formula: (C₂H₅)₂NCSSAg
Punto di fusione: 172…175 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004929
CAS Number: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9,III

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR