4,4'-Diaminostilbene-2,2'-disulphonic Acid ≥94.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Amsonic acid, 4,4-Diamino-2,2-stilbenedisulphonic acid
D0121-25G
D0121-500G
D0121-100G
TCIAD0121-25GEA
20
EUR
TCIAD0121-25G
TCIAD0121-500G
TCIAD0121-100G
4,4'-Diaminostilbene-2,2'-disulphonic Acid ≥94.0% (by titrimetric analysis)
4,4'-Diaminostilbene-2,2'-disulphonic Acid
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (Neutralization titration) | min. 94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...