Stampa…

4,4'-Diaminostilbene dihydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D0120-1G D0120-5G
TCIAD0120-1GEA 83 EUR
TCIAD0120-1G TCIAD0120-5G
4,4'-Diaminostilbene dihydrochloride ≥98.0% (by HPLC, titration analysis)
4,4'-Diaminostilbene dihydrochloride
Formula: C₁₄H₁₄N₂
MDL Number: MFCD00012984
CAS Number: 54760-75-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in hot 0.1M HCl almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR