Stampa…

DL-2,3-Diaminopropionic acid hydrobromide ≥98.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: 3-Amino-DL-alanine Hydrobromide, 2,3-Diaminopropanoic acid hydrobromide, H-DL-Dapa-OH·HBr

D0115-25G D0115-5G
TCIAD0115-25GEA 324 EUR
TCIAD0115-25G TCIAD0115-5G
DL-2,3-Diaminopropionic acid hydrobromide ≥98.0% (by titrimetric analysis)
DL-2,3-Diaminopropionic acid hydrobromide
Formula: C₃H₈N₂O₂
MDL Number: MFCD00012883
CAS Number: 18635-45-5
EINECS: 242-467-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR