2,4-Diaminodiphenylamine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N1-Phenylbenzene-1,2,4-triamine
D0087-25G
D0087-5G
TCIAD0087-25GEA
318
EUR
TCIAD0087-25G
TCIAD0087-5G
2,4-Diaminodiphenylamine ≥98.0% (by titrimetric analysis)
2,4-Diaminodiphenylamine
Formula:
C₁₂H₁₃N₃ Peso molecolare: 199,26 g/mol |
MDL Number:
MFCD00025285 CAS Number: 136-17-4 |
Risultati del test di specificazione
Appearance | White to Amber to Dark purple powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...