Stampa…

N,N'-Diacetyl-1,4-phenylenediamine ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: DAPPD, N,N'-1,4-Phenylenebisacetamide, N,N'-(1,4-Phenylene)diacetamide

D0068-25G D0068-5G
TCIAD0068-25GEA 160 EUR
TCIAD0068-25G TCIAD0068-5G
N,N'-Diacetyl-1,4-phenylenediamine ≥98.0% (by GC, total nitrogen)
N,N'-Diacetyl-1,4-phenylenediamine
Formula: C₁₀H₁₂N₂O₂
Peso molecolare: 192,22 g/mol
MDL Number: MFCD00026142
CAS Number: 140-50-1
EINECS: 205-417-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Green to Brown powder to crystaline
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Acetic acid almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR