Stampa…

4,4'-Diaminodiphenylamine sulphate Hydrate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N1-(4-Aminophenyl)benzene-1,4-diamine sulphate, Bis(4-aminophenyl)amine Sulphate, 4,4'-Iminodianiline sulphate

D0041-25G D0041-500G
TCIAD0041-25GEA 42 EUR
TCIAD0041-25G TCIAD0041-500G
4,4'-Diaminodiphenylamine sulphate Hydrate ≥97.0% (by titrimetric analysis)
4,4'-Diaminodiphenylamine sulphate Hydrate
Formula: C₁₂H₁₃N₃
Peso molecolare: 297,33 g/mol
MDL Number: MFCD00013113
CAS Number: 53760-27-3
EINECS: 258-748-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Dark blue powder to crystal
Purity (Neutralization titration) min. 97.0 % (after drying)
Drying loss 4.0 to 12.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR