Stampa…

4-Chloroquinoline 1-Oxide ≥97.0% (by GC, titration analysis)

Fornitore: TCI

C3682-1G C3682-250MG
TCIAC3682-1GEA 229 EUR
TCIAC3682-1G TCIAC3682-250MG
4-Chloroquinoline 1-Oxide ≥97.0% (by GC, titration analysis)
4-Chloroquinoline 1-Oxide
Formula: C₉H₆ClNO
MDL Number: MFCD18377747
CAS Number: 4637-59-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR