Stampa…

Cephalotaxine ≥98.0% (by GC)

Fornitore: TCI

C3641-100MG C3641-25MG
TCIAC3641-100MGEA 178 EUR
TCIAC3641-100MG TCIAC3641-25MG
Cephalotaxine ≥98.0% (by GC)
Cephalotaxine
Formula: C₁₈H₂₁NO₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD21090383
CAS Number: 24316-19-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -91 to -101 deg (C=0.5, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR