Stampa…

8-Chloro-1-(3-chloropropyl)theobromine ≥95.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 8-Chloro-1-(3-chloropropyl)-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione

C3618-5G C3618-25G
TCIAC3618-5GEA 112 EUR
TCIAC3618-5G TCIAC3618-25G
8-Chloro-1-(3-chloropropyl)theobromine ≥95.0% (by HPLC)
8-Chloro-1-(3-chloropropyl)theobromine

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR