2-Chlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Formula:
C₁₂H₁₆BClO₂ |
MDL Number:
MFCD05663846 CAS Number: 870195-94-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | max. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...