Stampa…

N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide Epoxide, N-(4-Cyano-3-(trifluoromethyl)phenyl)-2-methyloxirane-2-carboxamide

C3607-5G C3607-25G
TCIAC3607-5GEA 28 EUR
TCIAC3607-5G TCIAC3607-25G
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide ≥95.0% (by GC)
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide
Formula: C₁₂H₉F₃N₂O₂
Peso molecolare: 270,21 g/mol
MDL Number: MFCD08460201
CAS Number: 90357-51-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR