SCH-58261 ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
SCH 58261, 2-(furan-2-yl)-7-phenethyl-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine, SCH58261, SCH-58261
C3599-25MG
C3599-5MG
TCIAC3599-25MGEA
430
EUR
TCIAC3599-25MG
TCIAC3599-5MG
SCH-58261 ≥98.0% (by HPLC)
SCH-58261
Formula:
C₁₈H₁₅N₇O Peso molecolare: 345,36 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00933778 CAS Number: 160098-96-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 221.0 to 225.0 deg-C |
Elemental analysis (Nitrogen) | 27.00 to 29.80 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...