2-Chloro-6-nitrobenzaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
C3593-1G
C3593-5G
TCIAC3593-1GEA
55
EUR
TCIAC3593-1G
TCIAC3593-5G
2-Chloro-6-nitrobenzaldehyde ≥98.0% (by GC)
2-Chloro-6-nitrobenzaldehyde
Formula:
C₇H₄ClNO₃ Peso molecolare: 185,57 g/mol |
MDL Number:
MFCD00007204 CAS Number: 6361-22-4 EINECS: 228-831-6 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...