Stampa…

(Cyclohexyl-CAAC)Rh(COD)Cl ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Chloro[2-(2,6-diisopropylphenyl)-3,3-dimethyl-2-azaspiro[4.5]dec-1-ylidene][1,2,5,6-eta-1,5-cyclooctadiene]rhodium

C3592-1G C3592-100MG
TCIAC3592-1GEA 767 EUR
TCIAC3592-1G TCIAC3592-100MG
(Cyclohexyl-CAAC)Rh(COD)Cl ≥98.0% (by titrimetric analysis)
(Cyclohexyl-CAAC)Rh(COD)Cl
Formula: C₃₁H₄₇ClNRh
Peso molecolare: 572,08 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 1801869-83-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR