(S)-(+)-2-Chloro-1-phenylethanol ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₉ClO Punto di ebollizione: 114°C |
MDL Number:
MFCD00077692 CAS Number: 70111-05-6 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...