Stampa…

(R)-(-)-2-Chloro-1-phenylethanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (R)-2-Chloro-1-phenylethanol

C3561-25G C3561-5G
TCIAC3561-25GEA 301 EUR
TCIAC3561-25G TCIAC3561-5G
(R)-(-)-2-Chloro-1-phenylethanol ≥98.0% (by GC)
(R)-(-)-2-Chloro-1-phenylethanol
Formula: C₈H₉ClO
Peso molecolare: 156,6095 g/mol
MDL Number: MFCD00077691
CAS Number: 56751-12-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR