Stampa…

5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde

C3559-1G C3559-5G
TCIAC3559-1GEA 55 EUR
TCIAC3559-1G TCIAC3559-5G
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Formula: C₉H₁₁ClN₂O
Peso molecolare: 220,65 g/mol
MDL Number: MFCD00100755
CAS Number: 947-95-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 143.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR