4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₉H₁₈FNO₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD11053363 CAS Number: 86786-89-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...