Cyanomethyl Methyl(phenyl)carbamodithioate ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
Synonyms:
Methyl(phenyl)carbamodithioic acid Cyanomethyl ester
Formula:
C₁₀H₁₀N₂S₂ Peso molecolare: 222.32 g/mol |
MDL Number:
MFCD16038113 CAS Number: 76926-16-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...