Stampa…

3-Chlorosalicylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C3522-1G C3522-5G
TCIAC3522-1GEA 35 EUR
TCIAC3522-1G TCIAC3522-5G
3-Chlorosalicylic acid ≥98.0% (by HPLC, titration analysis)
3-Chlorosalicylic acid
Formula: C₇H₅ClO₃
Peso molecolare: 172,57 g/mol
MDL Number: MFCD00010390
CAS Number: 1829-32-9
EINECS: 217-379-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR