2-Chloro-4,6-di-p-tolyl-1,3,5-triazine ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₇H₁₄ClN₃ |
MDL Number:
MFCD29071841 CAS Number: 21902-34-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 205.0 to 209.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...