4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Cyano-3,5-difluorophenyl 4-Amylbenzoate, 4-Cyano-3,5-difluorophenyl 4-Pentylbenzoate, 4-Amylbenzoic acid 4-Cyano-3,5-difluorophenyl ester, 4-Pentylbenzoic acid 4-Cyano-3,5-difluorophenyl ester
C3512-1G
C3512-5G
TCIAC3512-1GEA
49
EUR
TCIAC3512-1G
TCIAC3512-5G
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate
Formula:
C₁₉H₁₇F₂NO₂ Peso molecolare: 329,347 g/mol |
MDL Number:
MFCD11053375 CAS Number: 123843-69-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...