Stampa…

Schisandrin A ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (6R,7S,12aR)-5,6,7,8-Tetrahydro-1,2,3,10,11,12-hexamethoxy-6,7-dimethyldibenzo[a,c]cyclooctene, Schizandrin A, Deoxyschizandrin, Deoxyschisandrin

C3501-100MG C3501-25MG
TCIAC3501-100MGEA 454 EUR
TCIAC3501-100MG TCIAC3501-25MG
Schisandrin A ≥98.0% (by GC)
Schisandrin A
Formula: C₂₄H₃₂O₆
Peso molecolare: 416,51 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09026934
CAS Number: 61281-38-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR