4-Chlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
C3500-25G
C3500-5G
TCIAC3500-25GEA
153
EUR
TCIAC3500-25G
TCIAC3500-5G
4-Chlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Chlorophenylboronic acid pinacol ester
Formula:
C₁₂H₁₆BClO₂ Peso molecolare: 238,52 g/mol |
MDL Number:
MFCD05663875 CAS Number: 195062-61-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...