Stampa…

4-Chloro-N-methoxy-N-methylbenzamide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C3486-5G C3486-25G
TCIAC3486-5GEA 53 EUR
TCIAC3486-5G TCIAC3486-25G
4-Chloro-N-methoxy-N-methylbenzamide ≥98.0% (by GC)
4-Chloro-N-methoxy-N-methylbenzamide
Formula: C₉H₁₀ClNO₂
Peso molecolare: 199,63 g/mol
MDL Number: MFCD02684308
CAS Number: 122334-37-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR