4-Chloro-3-iodobenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
C3484-1G
C3484-5G
TCIAC3484-1GEA
20
EUR
TCIAC3484-1G
TCIAC3484-5G
4-Chloro-3-iodobenzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-3-iodobenzoic acid
Formula:
C₇H₄ClIO₂ |
MDL Number:
MFCD00079732 CAS Number: 42860-04-8 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 216.0 to 220.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...