Stampa…

4-(9H-Carbazol-9-yl)benzaldehyde ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-(4-Formylphenyl)-9H-carbazole

C3481-1G C3481-5G
TCIAC3481-1GEA 62 EUR
TCIAC3481-1G TCIAC3481-5G
4-(9H-Carbazol-9-yl)benzaldehyde ≥96.0% (by GC)
4-(9H-Carbazol-9-yl)benzaldehyde
Formula: C₁₉H₁₃NO
Peso molecolare: 271,32 g/mol
MDL Number: MFCD19441374
CAS Number: 110677-45-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Amber powder to crystal
Purity (GC) min. 96.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR