Stampa…

Columbianadin ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 2-[(8S)-2-oxo-8,9-dihydrofuro[2,3-h]chromen-8-yl]propan-2-yl (Z)-2-methylbut-2-enoate

C3479-100MG C3479-25MG
TCIAC3479-100MGEA 758 EUR
TCIAC3479-100MG TCIAC3479-25MG
Columbianadin ≥98.0% (by HPLC)
Columbianadin
Formula: C₁₉H₂₀O₅
Peso molecolare: 328,36 g/mol
MDL Number: MFCD07781426
CAS Number: 5058-13-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 68.00 to 71.00 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR