Stampa…

Carnosic acid ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic acid

C3477-20MG C3477-100MG
TCIAC3477-20MGEA 55 EUR
TCIAC3477-20MG TCIAC3477-100MG
Carnosic acid ≥97.0% (by HPLC, titration analysis)
Carnosic acid
Formula: C₂₀H₂₈O₄
Peso molecolare: 332.44 g/mol
Punto di fusione: 190°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD02259459
CAS Number: 3650-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e17a6ae9b1caa68c02e2d0baa1783cfe85c99f8e&vid=e17a6ae9b1caa68c02e2d0baa1783cfe85c99f8e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +182 to +195 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR