FCCP ≥95.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
FCCP, [4-(Trifluoromethoxy)phenyl]carbonohydrazonoyl dicyanide, [4-(Trifluoromethoxy)phenylhydrazono]malononitrile, Carbonyl cyanide 4-(Trifluoromethoxy)phenylhydrazone
Formula:
C₁₀H₅F₃N₄O |
MDL Number:
MFCD00009699 CAS Number: 370-86-5 EINECS: 206-730-8 UN: 3439 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...