Stampa…

Cinobufagin ≥98.0% (by HPLC)

Fornitore: TCI
Pericolo

C3460-100MG C3460-25MG
TCIAC3460-100MGEA 504 EUR
TCIAC3460-100MG TCIAC3460-25MG
Cinobufagin ≥98.0% (by HPLC)
Cinobufagin
Formula: C₂₆H₃₄O₆
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00058625
CAS Number: 470-37-1
UN: 2811
ADR: 6.1,I

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 213.0 to 217.0 deg-C
Specific rotation [a]20/D -3.0 to -4.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR