Stampa…

Cinnarizine ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: 1-Cinnamyl-4-diphenylmethylpiperazine, 1-(Diphenylmethyl)-4-(3-phenyl-2-propenyl)piperazine, 1-Benzhydryl-4-cinnamylpiperazine

C3459-10G C3459-50G
TCIAC3459-10GEA 42 EUR
TCIAC3459-10G TCIAC3459-50G
Cinnarizine ≥98.0% (by HPLC, total nitrogen)
Cinnarizine
Formula: C₂₆H₂₈N₂
Peso molecolare: 368,524 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00056037
CAS Number: 298-57-7
EINECS: 206-064-8
Merck Index: 14,02305
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d81ff6a23d6973ad4ab1d5d684e0c358d1609275&vid=d81ff6a23d6973ad4ab1d5d684e0c358d1609275&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR