Stampa…

Chloro(methyl)(phenyl)(vinyl)silane ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

C3434-5G C3434-25G
TCIAC3434-5GEA 62 EUR
TCIAC3434-5G TCIAC3434-25G
Chloro(methyl)(phenyl)(vinyl)silane ≥98.0% (by GC)
Chloro(methyl)(phenyl)(vinyl)silane
Formula: C₉H₁₁ClSi
Peso molecolare: 182,72 g/mol
Punto di ebollizione: 80°C
Flash Pt: 72°C
MDL Number: MFCD00053208
CAS Number: 17306-05-7
EINECS: 241-333-3
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5419327626fb9344cf511894aa2e3bca0a97635c&vid=5419327626fb9344cf511894aa2e3bca0a97635c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR