Cyclothiazide ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
6-Chloro-3,4-dihydro-3-(2-norbornen-5-yl)-2H-1,2-4-benzothiadiazine-7-sulphonamide 1,1-Dioxide, Cyclothiazide (mixture of isomers)
C3392-100MG
C3392-25MG
TCIAC3392-100MGEA
209
EUR
TCIAC3392-100MG
TCIAC3392-25MG
Cyclothiazide ≥95.0% (by HPLC)
Cyclothiazide
Formula:
C₁₄H₁₆ClN₃O₄S₂ Peso molecolare: 389,87 g/mol Punto di fusione: 234°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00210192 CAS Number: 2259-96-3 EINECS: 218-859-7 Merck Index: 14,02754 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 10.20 to 11.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...